Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006141.hg.1 |
ENST00000319441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006141.hg.1 |
ENST00000535860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006141.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006141.hg.1 |
TC20000450.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006135.hg.1 |
ENST00000319441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006135.hg.1 |
ENST00000535860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006135.hg.1 |
ENST00000543666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006135.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006135.hg.1 |
TC20000450.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006137.hg.1 |
ENST00000319441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006137.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006137.hg.1 |
TC20000450.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
TC2000007864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154603.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
TC2000007864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154607.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000319441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000467047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000470051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000475833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000485958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
ENST00000498194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007864.hg.1 |
NM_002591.3 |
Refseq |