Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006782.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006782.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006782.hg.1 |
TC16000487.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007997.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158635.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158635.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158635.hg.1 |
ENST00000219235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158635.hg.1 |
NM_002990.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158635.hg.1 |
TC1600007997.hg.1 |
Clariom D TC |