Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015998.hg.1 |
ENST00000262498.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015998.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015998.hg.1 |
TC16001147.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180535.hg.1 |
TC1600010475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180536.hg.1 |
TC1600010475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000262498.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000562622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000564150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000565880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
ENST00000567660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010475.hg.1 |
NM_013242.2 |
Refseq |