Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012163.hg.1 |
TC06001035.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012163.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012163.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012162.hg.1 |
TC06001035.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012162.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012162.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165988.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165988.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165988.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165988.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165988.hg.1 |
TC0600009628.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009628.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165986.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165986.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165986.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165986.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165986.hg.1 |
TC0600009628.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600165987.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165987.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165987.hg.1 |
ENST00000367658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165987.hg.1 |
NM_016217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165987.hg.1 |
TC0600009628.hg.1 |
Clariom D TC |