Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002019.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002019.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002019.hg.1 |
TC20000162.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002017.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002017.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002017.hg.1 |
TC20000162.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002018.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002018.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002018.hg.1 |
TC20000162.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000482637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
ENST00000482637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147719.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147719.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147719.hg.1 |
TC2000006993.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147719.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147719.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147723.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147723.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147723.hg.1 |
TC2000006993.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147723.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147723.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000482637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
ENST00000482637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006993.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147718.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147718.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147718.hg.1 |
TC2000006993.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147718.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147718.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147722.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147722.hg.1 |
NM_024893.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147722.hg.1 |
TC2000006993.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147722.hg.1 |
ENST00000376862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147722.hg.1 |
NM_024893.2 |
Refseq |