Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007669.hg.1 |
TC06000631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007669.hg.1 |
ENST00000230588.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007669.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157186.hg.1 |
TC0600008169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000230588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
ENST00000611727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008169.hg.1 |
NM_005588.2 |
Refseq |