Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
TC10002950.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
ENST00000415747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
ENST00000423766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
ENST00000433452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
ENST00000458230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
ENST00000463201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011799.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186408.hg.1 |
TC1000012511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000433452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
ENST00000463201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_001145201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012511.hg.1 |
NM_145202.4 |
Refseq |