Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000558569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000558569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000558569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000560707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000560707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
ENST00000560707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000240587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000558569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000558569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000558569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000560707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000560707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
ENST00000560707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010334.hg.1 |
NM_020856.3 |
Refseq |