Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025233.hg.1 |
TC03001415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025233.hg.1 |
ENST00000480687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025233.hg.1 |
ENST00000535833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025233.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025231.hg.1 |
TC03001415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025231.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025231.hg.1 |
ENST00000480687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025231.hg.1 |
ENST00000535833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025231.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025232.hg.1 |
TC03001415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025232.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025232.hg.1 |
ENST00000480687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025232.hg.1 |
ENST00000535833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025232.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207636.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207636.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207636.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207636.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207636.hg.1 |
TC0300013979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300207638.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207638.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207638.hg.1 |
TC0300013979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300207635.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207635.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207635.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207635.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207635.hg.1 |
TC0300013979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300207637.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207637.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207637.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207637.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207637.hg.1 |
TC0300013979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
ENST00000320431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013979.hg.1 |
NM_198722.2 |
Refseq |