Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023945.2 | NP_076434.2 | 2 - 3 | 364 | 258 | |
GenBank mRNA | AB013103.1 | - | 2 - 3 | 329 | 258 | |
AF237907.1 | - | 2 - 3 | 332 | 258 | ||
AF321127.1 | - | 2 - 3 | 334 | 258 | ||
BC029884.1 | - | 2 - 3 | 364 | 5171526 | 258 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |