Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284510.1 | NP_001271439.1 | 1 - 2 | 187 | 124 | |
GenBank mRNA | AJ489980.1 | - | 1 - 2 | 99 | 124 | |
AK297612.1 | - | 1 - 2 | 132 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Homo sapiens Probe spans exons |