Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252043.1 | NP_001238972.1 | 14 - 15 | 2607 | 179 | |
NM_004783.3 | NP_004774.1 | 14 - 15 | 2607 | 179 | ||
NM_016151.3 | NP_057235.2 | 14 - 15 | 2607 | 179 | ||
XM_011545982.1 | XP_011544284.1 | 14 - 15 | 2854 | 179 | ||
XM_011545983.2 | XP_011544285.1 | 14 - 15 | 2851 | 179 | ||
XM_011545984.1 | XP_011544286.1 | 14 - 15 | 2854 | 179 | ||
XM_011545985.1 | XP_011544287.1 | 14 - 15 | 2851 | 179 | ||
XM_011545986.2 | XP_011544288.1 | 14 - 15 | 2854 | 179 | ||
GenBank mRNA | AB020688.1 | - | 14 - 15 | 2193 | 179 | |
AF061943.1 | - | 14 - 15 | 2548 | 179 | ||
AF263313.1 | - | 13 - 14 | 1790 | 179 | ||
AK291473.1 | - | 14 - 15 | 2124 | 179 | ||
AK304446.1 | - | 6 - 7 | 1113 | 179 | ||
AL137701.1 | - | 6 - 7 | 2444 | 179 | ||
BC015204.1 | - | 14 - 15 | 2573 | 179 | ||
BC051798.1 | - | 14 - 15 | 2364 | 179 | ||
BC054495.1 | - | 14 - 15 | 2560 | 179 | ||
BC136653.1 | - | 14 - 15 | 2292 | 9020643 | 179 | |
BC136655.1 | - | 14 - 15 | 2292 | 9020645 | 179 | |
BC142663.1 | - | 14 - 15 | 1947 | 40148112 | 179 | |
BC144344.1 | - | 14 - 15 | 2292 | 9052864 | 179 | |
BC151221.1 | - | 14 - 15 | 1947 | 40148752 | 179 | |
BC152413.1 | - | 14 - 15 | 2193 | 8862518 | 179 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |