Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000029.3 | NP_000020.1 | 2 - 3 | 1363 | 171 | |
GenBank mRNA | AB209769.1 | - | 2 - 3 | 875 | 171 | |
AK222798.1 | - | 2 - 3 | 894 | 171 | ||
AK222815.1 | - | 2 - 3 | 895 | 171 | ||
AK293507.1 | - | 3 - 4 | 648 | 171 | ||
AK307978.1 | - | 3 - 4 | 809 | 171 | ||
AK312291.1 | - | 2 - 3 | 973 | 171 | ||
BC011519.1 | - | 2 - 3 | 883 | 4213559 | 171 | |
BT006851.1 | - | 1 - 2 | 855 | 171 | ||
K02215.1 | - | 2 - 3 | 894 | 171 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |