Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012334.2 | NP_036466.2 | 30 - 31 | 4508 | 132 | |
XM_005248306.4 | XP_005248363.1 | 12 - 13 | 3402 | 132 | ||
XM_005248307.2 | XP_005248364.1 | 12 - 13 | 2878 | 132 | ||
XM_006714475.2 | XP_006714538.1 | 29 - 30 | 4215 | 132 | ||
XM_011514046.2 | XP_011512348.1 | 12 - 13 | 2581 | 132 | ||
GenBank mRNA | AB018342.2 | - | 30 - 31 | 4495 | 132 | |
AF132021.1 | - | 30 - 31 | 4120 | 132 | ||
AF234532.1 | - | 30 - 31 | 4262 | 132 | ||
AF247457.2 | - | 30 - 31 | 4193 | 132 | ||
AK074717.1 | - | 6 - 7 | 854 | 132 | ||
AK295630.1 | - | 5 - 6 | 756 | 132 | ||
AK295736.1 | - | 12 - 13 | 2363 | 132 | ||
AK316138.1 | - | 12 - 13 | 2536 | 132 | ||
AL832428.1 | - | 20 - 21 | 2962 | 129 | ||
BC137168.1 | - | 30 - 31 | 4217 | 9021165 | 132 | |
BC150285.1 | - | 30 - 31 | 4495 | 8860110 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |