Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015710.hg.1 |
ENST00000631284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018005.hg.1 |
TC01006326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018005.hg.1 |
ENST00000331380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018005.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009880.hg.1 |
ENST00000331380.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009880.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009880.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012212.hg.1 |
ENST00000456007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012212.hg.1 |
ENST00000456007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012212.hg.1 |
ENST00000456007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012212.hg.1 |
ENST00000456007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009880.hg.1 |
ENST00000331380.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009880.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009880.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015710.hg.1 |
ENST00000631284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171262.hg.1 |
TC0100009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100171263.hg.1 |
ENST00000331380.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171263.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171263.hg.1 |
ENST00000331380.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171263.hg.1 |
NM_003517.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171263.hg.1 |
TC0100009880.hg.1 |
Clariom D TC |