Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199208.1 | NP_001186137.1 | 3 - 4 | 390 | 95 | |
NM_001199209.1 | NP_001186138.1 | 3 - 4 | 374 | 95 | ||
XM_017026814.1 | XP_016882303.1 | 2 - 3 | 468 | 95 | ||
XM_017026815.1 | XP_016882304.1 | 2 - 3 | 473 | 95 | ||
GenBank mRNA | AF054821.1 | - | 2 - 3 | 371 | 95 | |
AK296802.1 | - | 3 - 4 | 389 | 95 | ||
AK304200.1 | - | 3 - 4 | 387 | 95 | ||
AK316136.1 | - | 3 - 4 | 374 | 95 | ||
BC136299.1 | - | 3 - 4 | 382 | 9020286 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection P450 Homo sapiens Inventoried Probe spans exons |