Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010239.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010239.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010239.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010239.hg.1 |
NM_001005184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175099.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175099.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175099.hg.1 |
TC0100010239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010239.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010239.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010239.hg.1 |
ENST00000368144.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010239.hg.1 |
NM_001005184.1 |
Refseq |