Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000780.hg.1 |
TC08000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000780.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000780.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006705.hg.1 |
ENST00000585729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006702.hg.1 |
ENST00000588001.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006702.hg.1 |
NM_001282300.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006702.hg.1 |
NM_001282300.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
NM_001102614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
NM_001102614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006750.hg.1 |
NM_001102614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000779.hg.1 |
TC08000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000779.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000779.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145644.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145644.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145644.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145644.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145644.hg.1 |
TC0800006739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145646.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145646.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145646.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145646.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145646.hg.1 |
TC0800006739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145645.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145645.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145645.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145645.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145645.hg.1 |
TC0800006739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1800006705.hg.1 |
ENST00000585729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
ENST00000382435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006739.hg.1 |
NM_054028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
ENST00000412468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
NM_001102614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
NM_001102614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006750.hg.1 |
NM_001102614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006702.hg.1 |
ENST00000588001.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006702.hg.1 |
NM_001282300.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006702.hg.1 |
NM_001282300.1 |
Refseq |