Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012702.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012702.hg.1 |
ENST00000535862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012702.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012702.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012698.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012698.hg.1 |
ENST00000535862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012698.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012698.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012706.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012706.hg.1 |
ENST00000535862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012706.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012706.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012701.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012701.hg.1 |
ENST00000535862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012701.hg.1 |
ENST00000557668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012701.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012701.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012016.hg.1 |
ENST00000502554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012708.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012708.hg.1 |
ENST00000535862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012708.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012708.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012705.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012705.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012705.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012704.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012704.hg.1 |
ENST00000535862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012704.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012704.hg.1 |
TC14001122.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
ENST00000557458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009134.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
ENST00000553731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167453.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400167462.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167462.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167462.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167462.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167462.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012016.hg.1 |
ENST00000502554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167452.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400167455.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167455.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167455.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167455.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167455.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167458.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
ENST00000555720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167454.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
ENST00000324679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
ENST00000556735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
NM_021818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167459.hg.1 |
TC1400009134.hg.1 |
Clariom D TC |