Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000397995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000554073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000554073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000554073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
ENST00000304704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
ENST00000336811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000304.hg.1 |
TC14000067.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144560.hg.1 |
TC1400006529.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000336811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000397995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000553909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000554073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000554073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000554073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
ENST00000555835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001145.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282192.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_001282193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_002937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006529.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
ENST00000397990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
ENST00000555597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
NM_001097577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
NM_194431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144558.hg.1 |
TC1400006529.hg.1 |
Clariom D TC |