Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252065.1 | NP_001238994.1 | 8 - 9 | 1189 | 179 | |
NM_001300773.1 | NP_001287702.1 | 8 - 9 | 1096 | 179 | ||
NM_004200.3 | NP_004191.2 | 7 - 8 | 964 | 179 | ||
XM_005274383.4 | XP_005274440.1 | 11 - 12 | 1851 | 179 | ||
XM_005274384.2 | XP_005274441.1 | 10 - 11 | 1855 | 179 | ||
XM_005274385.4 | XP_005274442.1 | 10 - 11 | 1621 | 179 | ||
XM_005274387.4 | XP_005274444.1 | 5 - 6 | 1888 | 179 | ||
XM_005274390.4 | XP_005274447.1 | 4 - 5 | 927 | 179 | ||
XM_006718736.3 | XP_006718799.1 | 4 - 5 | 898 | 179 | ||
XM_011545335.2 | XP_011543637.1 | 11 - 12 | 1817 | 179 | ||
XM_011545336.2 | XP_011543638.1 | 10 - 11 | 1692 | 179 | ||
XM_011545337.2 | XP_011543639.1 | 10 - 11 | 1684 | 179 | ||
XM_011545338.2 | XP_011543640.1 | 10 - 11 | 1589 | 179 | ||
XM_011545339.2 | XP_011543641.1 | 9 - 10 | 1316 | 179 | ||
XM_011545340.2 | XP_011543642.1 | 8 - 9 | 1182 | 179 | ||
XM_011545341.2 | XP_011543643.1 | 8 - 9 | 1086 | 179 | ||
XM_011545343.2 | XP_011543645.1 | 7 - 8 | 950 | 179 | ||
GenBank mRNA | AF038535.1 | - | 7 - 8 | 957 | 179 | |
BC125170.1 | - | 7 - 8 | 922 | 40121712 | 179 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |