Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011528504.1 | XP_011526806.1 | 4 - 5 | 1451 | 146 | |
XM_011528505.1 | XP_011526807.1 | 4 - 5 | 1518 | 146 | ||
XM_011528506.1 | XP_011526808.1 | 4 - 5 | 8302 | 146 | ||
XM_011528507.1 | XP_011526809.1 | 4 - 5 | 1386 | 146 | ||
XM_011528508.1 | XP_011526810.1 | 4 - 5 | 1527 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |