Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01590971_m1
See other MBNL1 GE Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01590971_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001314057.1
NP_001300986.1
10 - 11
1776
198
NM_021038.4
NP_066368.2
8 - 9
2507
198
NM_207292.2
NP_997175.1
7 - 8
2471
162
NM_207293.1
NP_997176.1
7 - 8
1900
162
NM_207294.1
NP_997177.1
5 - 6
1642
162
NM_207295.1
NP_997178.1
6 - 7
1678
198
XM_005247457.4
XP_005247514.1
10 - 11
2129
198
XM_005247458.4
XP_005247515.1
10 - 11
3383
198
XM_005247460.4
XP_005247517.1
10 - 11
2092
198
XM_005247461.4
XP_005247518.1
9 - 10
2093
162
XM_005247462.4
XP_005247519.1
9 - 10
2075
198
XM_005247463.4
XP_005247520.1
8 - 9
2039
162
XM_005247464.4
XP_005247521.1
9 - 10
2187
198
XM_005247466.4
XP_005247523.1
8 - 9
2128
162
XM_005247474.3
XP_005247531.1
8 - 9
1047
198
XM_005247475.3
XP_005247532.1
9 - 10
1370
198
XM_005247476.4
XP_005247533.1
7 - 8
1083
198
XM_005247477.4
XP_005247534.1
8 - 9
1136
198
XM_006713639.1
XP_006713702.1
10 - 11
2111
198
XM_011512843.2
XP_011511145.1
9 - 10
1925
198
XM_011512845.2
XP_011511147.1
8 - 9
1871
198
XM_011512846.2
XP_011511148.1
7 - 8
1835
162
XM_011512847.2
XP_011511149.1
7 - 8
1834
198
XM_011512848.2
XP_011511150.1
10 - 11
1733
198
XM_011512849.2
XP_011511151.1
6 - 7
1742
162
XM_017006423.1
XP_016861912.1
9 - 10
2380
198
XM_017006424.1
XP_016861913.1
9 - 10
2207
198
XM_017006425.1
XP_016861914.1
8 - 9
2173
162
XM_017006426.1
XP_016861915.1
8 - 9
2171
162
XM_017006427.1
XP_016861916.1
8 - 9
2300
198
XM_017006428.1
XP_016861917.1
8 - 9
1964
198
XM_017006429.1
XP_016861918.1
8 - 9
2047
198
XM_017006430.1
XP_016861919.1
7 - 8
2075
162
XM_017006431.1
XP_016861920.1
7 - 8
2152
162
XM_017006435.1
XP_016861924.1
9 - 10
1177
198
XM_017006438.1
XP_016861927.1
7 - 8
1796
162
XM_017006439.1
XP_016861928.1
8 - 9
1316
198
XM_017006440.1
XP_016861929.1
7 - 8
1050
198
XM_017006441.1
XP_016861930.1
6 - 7
1047
162
XM_017006442.1
XP_016861931.1
6 - 7
1150
162
GenBank mRNA
AB007888.1
-
7 - 8
2471
162
AF255334.1
-
5 - 6
1364
162
AF395876.1
-
6 - 7
889
198
AF401998.1
-
7 - 8
1093
198
AJ308400.1
-
7 - 8
1122
198
AK297133.1
-
4 - 5
562
162
AK304089.1
-
7 - 8
856
198
BC043493.1
-
8 - 9
2012
6044671
198
BC050535.1
-
10 - 11
1297
6042311
198
Y13829.1
-
7 - 8
1622
162
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene MBNL1
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0