Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021048.hg.1 |
ENST00000361198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021048.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021048.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021048.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021048.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021048.hg.1 |
TC10001617.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021047.hg.1 |
ENST00000361198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021047.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021047.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021047.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021047.hg.1 |
TC10001617.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179873.hg.1 |
TC1000011669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
ENST00000490751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011669.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
ENST00000361198.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
ENST00000425280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
ENST00000461873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
NM_001113407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
NM_003893.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179872.hg.1 |
TC1000011669.hg.1 |
Clariom D TC |