Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006465.hg.1 |
TC16002034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006465.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006465.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007731.hg.1 |
ENST00000262499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006466.hg.1 |
TC16002034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006466.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006466.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158037.hg.1 |
TC1600007957.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007731.hg.1 |
ENST00000262499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158035.hg.1 |
TC1600007957.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000245185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000561491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000562017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000563985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
ENST00000567300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007957.hg.1 |
NM_005953.4 |
Refseq |