Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000990.4 | NP_000981.1 | 5 | 4463 | 87 | |
GenBank mRNA | AK125453.1 | - | 1 | 2413 | 87 | |
BC004284.1 | - | 1 | 1775 | 87 | ||
BC014919.1 | - | 1 | 889 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |