Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000423958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000457545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
ENST00000467156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009231.hg.1 |
NM_001080538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000519451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010116.hg.1 |
ENST00000524132.1 |
Ensembl |