Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009016.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009016.hg.1 |
ENST00000548757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009016.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009016.hg.1 |
TC12000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009013.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009013.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009013.hg.1 |
TC12000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009014.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009014.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009014.hg.1 |
TC12000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009015.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009015.hg.1 |
ENST00000548757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009015.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009015.hg.1 |
TC12000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008302.hg.1 |
ENST00000467527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008302.hg.1 |
ENST00000467527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008302.hg.1 |
ENST00000467527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008302.hg.1 |
ENST00000467527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161517.hg.1 |
TC1200008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000547468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000548511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000548757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000549345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000547468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000548511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000548757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000549345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161523.hg.1 |
TC1200008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161512.hg.1 |
TC1200008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161516.hg.1 |
TC1200008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000548511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000548757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000549345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000548511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000548757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000549345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161521.hg.1 |
TC1200008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000356891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000453037.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000550333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000552121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000552803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
ENST00000552847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
NM_001009894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161518.hg.1 |
TC1200008370.hg.1 |
Clariom D TC |