Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032732.hg.1 |
TC02002045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032732.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032732.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032732.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032731.hg.1 |
TC02002045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032731.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032731.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032731.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032727.hg.1 |
TC02002045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032727.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032727.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032727.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032727.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032729.hg.1 |
TC02002045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032729.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032729.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032729.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197042.hg.1 |
TC0200013284.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197040.hg.1 |
TC0200013284.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197044.hg.1 |
TC0200013284.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197046.hg.1 |
TC0200013284.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000420686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
ENST00000478626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013284.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
ENST00000306336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
ENST00000409734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
ENST00000423181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
NM_001013649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197045.hg.1 |
TC0200013284.hg.1 |
Clariom D TC |