Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009399.hg.1 |
TC0X000730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009399.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009399.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007353.hg.1 |
ENST00000431286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009398.hg.1 |
TC0X000730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009398.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009398.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007353.hg.1 |
ENST00000431286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000370361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
ENST00000477649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008731.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159305.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159305.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159305.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159305.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159305.hg.1 |
TC0X00008731.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159306.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159306.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159306.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159306.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159306.hg.1 |
TC0X00008731.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159307.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159307.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159307.hg.1 |
ENST00000330374.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159307.hg.1 |
NM_001017980.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159307.hg.1 |
TC0X00008731.hg.1 |
Clariom D TC |