Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007709.hg.1 |
TC10002972.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007709.hg.1 |
ENST00000285979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007709.hg.1 |
ENST00000339022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007709.hg.1 |
ENST00000476630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007709.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007709.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007708.hg.1 |
TC10002972.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007708.hg.1 |
ENST00000285979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007708.hg.1 |
ENST00000339022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007708.hg.1 |
ENST00000476630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007708.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007708.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007706.hg.1 |
TC10002972.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007706.hg.1 |
ENST00000285979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007706.hg.1 |
ENST00000339022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007706.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007706.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007705.hg.1 |
TC10002972.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007705.hg.1 |
ENST00000285979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007705.hg.1 |
ENST00000339022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007705.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007705.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185950.hg.1 |
TC1000012480.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185951.hg.1 |
TC1000012480.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185952.hg.1 |
TC1000012480.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012480.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
ENST00000285979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
ENST00000339022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
NM_000772.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
NM_001128925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185949.hg.1 |
TC1000012480.hg.1 |
Clariom D TC |