Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
TC06002209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
ENST00000389942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
ENST00000409806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
ENST00000416573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
ENST00000542614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027668.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193287.hg.1 |
TC0600013519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |