Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080410.2 | NP_001073879.2 | 8 - 9 | 2177 | 94 | |
XM_005264158.3 | XP_005264215.1 | 6 - 7 | 1096 | 94 | ||
XM_006711950.2 | XP_006712013.1 | 13 - 14 | 3161 | 94 | ||
XM_011532562.1 | XP_011530864.1 | 9 - 10 | 2609 | 94 | ||
XM_011532563.2 | XP_011530865.1 | 7 - 8 | 1534 | 94 | ||
XM_017003421.1 | XP_016858910.1 | 10 - 11 | 5258 | 94 | ||
XM_017003422.1 | XP_016858911.1 | 13 - 14 | 4903 | 94 | ||
XM_017003423.1 | XP_016858912.1 | 13 - 14 | 4284 | 94 | ||
GenBank mRNA | AB075820.1 | - | 8 - 9 | 2015 | 94 | |
AK127503.1 | - | 5 - 6 | 658 | 94 | ||
BC112202.1 | - | 7 - 8 | 1085 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons |