Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000341040.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000406721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000474369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000494294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000497771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000527229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000529013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
ENST00000532006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012288.hg.1 |
NM_001001712.2 |
Refseq |