Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145030.1 | NP_001138502.1 | 7 - 8 | 3409 | 129 | |
XM_011533694.2 | XP_011531996.1 | 7 - 8 | 3409 | 129 | ||
XM_017006361.1 | XP_016861850.1 | 7 - 8 | 3409 | 129 | ||
XM_017006362.1 | XP_016861851.1 | 7 - 8 | 3409 | 129 | ||
GenBank mRNA | BX648094.1 | - | 6 - 7 | 1911 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |