Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004706.hg.1 |
TC03000235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004706.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004706.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150782.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150782.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150782.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150782.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150782.hg.1 |
TC0300007205.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
ENST00000309765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007205.hg.1 |
NM_001145030.1 |
Refseq |