Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105529.1 | NP_001098999.1 | 33 - 34 | 3403 | 110 | |
NM_006095.2 | NP_006086.1 | 34 - 35 | 3448 | 110 | ||
XM_005248043.2 | XP_005248100.1 | 34 - 35 | 3443 | 110 | ||
XM_011513615.1 | XP_011511917.1 | 35 - 36 | 3464 | 110 | ||
XM_011513616.2 | XP_011511918.1 | 35 - 36 | 3498 | 110 | ||
XM_011513618.1 | XP_011511920.1 | 34 - 35 | 3419 | 110 | ||
XM_017007645.1 | XP_016863134.1 | 33 - 34 | 3432 | 110 | ||
XM_017007646.1 | XP_016863135.1 | 34 - 35 | 3371 | 110 | ||
XM_017007647.1 | XP_016863136.1 | 32 - 33 | 3303 | 110 | ||
GenBank mRNA | AB013452.1 | - | 34 - 35 | 3229 | 110 | |
AB209687.1 | - | 33 - 34 | 3256 | 110 | ||
AF067820.1 | - | 34 - 35 | 3216 | 110 | ||
BC109317.1 | - | 33 - 34 | 3165 | 40036536 | 110 | |
BC109318.1 | - | 33 - 34 | 3171 | 40036538 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons |