Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000478817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000478817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019794.hg.1 |
TC09001578.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019794.hg.1 |
ENST00000259457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019794.hg.1 |
ENST00000498485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019794.hg.1 |
ENST00000536392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019794.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019798.hg.1 |
TC09001578.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019798.hg.1 |
ENST00000259457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019798.hg.1 |
ENST00000536392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019798.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019793.hg.1 |
TC09001578.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019793.hg.1 |
ENST00000259457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019793.hg.1 |
ENST00000498485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019793.hg.1 |
ENST00000536392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019793.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178349.hg.1 |
TC0900011496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000474081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000478817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000478817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011496.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178344.hg.1 |
TC0900011496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000259457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000441097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000466951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
ENST00000498485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178345.hg.1 |
TC0900011496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900178353.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178353.hg.1 |
NM_002799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178353.hg.1 |
TC0900011496.hg.1 |
Clariom D TC |