Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000447.2 | NP_000438.2 | 11 - 12 | 1498 | 154 | |
XM_005273199.3 | XP_005273256.1 | 10 - 11 | 1311 | 154 | ||
XM_017001835.1 | XP_016857324.1 | 10 - 11 | 1819 | 154 | ||
GenBank mRNA | AF416718.1 | - | 9 - 10 | 1139 | 154 | |
AK301143.1 | - | 10 - 11 | 1247 | 154 | ||
BC006365.2 | - | 12 - 13 | 1692 | 4131408 | 154 | |
BT006984.1 | - | 9 - 10 | 1071 | 154 | ||
L43964.1 | - | 10 - 11 | 1438 | 154 | ||
L44577.1 | - | 11 - 12 | 1436 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |