Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023013.4 | NP_075389.2 | 2 | 260 | 153 | |
GenBank mRNA | AK125568.1 | - | 2 | 260 | 153 | |
AL049686.1 | - | 2 | 260 | 153 | ||
BC144199.1 | - | 2 | 230 | 9052719 | 153 | |
DA903707.1 | - | 2 | 260 | 153 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291380.1 | NP_001278309.1 | 2 | 153 |
Gene Symbol: PRAMEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024661.2 | NP_001019832.2 | 2 | 153 |
Set Membership: |
Detect Genomic DNA Both primers and probe map within a single exon |