Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005269006.3 | XP_005269063.1 | 5 | 1231 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |