Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018111.2 | NP_060581.2 | 1 | 654 | 63 | |
GenBank mRNA | AK001352.1 | - | 1 | 654 | 63 | |
BC051004.1 | - | 1 | 659 | 6483373 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17152187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10985597 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10985599 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3361442 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P153524 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P48522 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P153524 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 14833 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 32648 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12885 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42985 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P21027190 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P153524 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3361442 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P153524 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3361442 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31623061 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10985598 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10985597 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10985599 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17152187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10985599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10985597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17152187 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10985597 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10985599 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17152187 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31623061 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10985598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10985599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17152187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10985597 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P153524 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P48522 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10985600 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10985597 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10985599 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10985601 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17152189 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10985599 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17152188 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17152190 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17152192 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10985600 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17152191 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P48522 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P153524 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985565 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985597 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985584 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985574 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152176 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985587 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985600 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152188 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985578 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985599 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152180 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152173 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985568 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985583 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152187 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985566 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152169 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152178 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17152181 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985580 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_018111_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038315 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038318 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038317 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038316 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8038314 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220151_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11725526_at | - | - |
Affymetrix Human Genome U219 Array | 11725526_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023335.hg.1 | ENST00000408991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023335.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023335.hg.1 | TC19001708.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011124.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900193104.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900193104.hg.1 | NM_018111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900193104.hg.1 | TC1900011124.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900193104.hg.1 | ENST00000408991.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900193104.hg.1 | NM_018111.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 220151_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 220151_at | - | - |
Human Genome U133A 2.0 Array | 220151_PM_at | - | - |
Human Genome U133A 2.0 Array | 220151_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_018111_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw570872 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220151_at5 | - | - |
Human Genome U95 Set | 80163_at | - | - |
Human Genome U95 Set | 80163_PM_at | - | - |
Human Genome U133 Set | 220151_PM_at | - | - |
Human Genome U133 Set | 220151_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011683 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011683 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 212606 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1721628 | AK001352.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1721628 | BC051004.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1721628 | BC069669.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922458-S | NM_018111.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922458-S | AK001352.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922458-S | BC051004.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922458-S | BC069669.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922458-S | BC069693.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_13670-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001410136 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001410136 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6840372 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13670 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1721628 | BC069693.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1721628 | NM_018111.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1721628 | AK001352.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1721628 | BC051004.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1721628 | BC069669.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1721628 | BC069693.1 | human_transcriptome |