Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015252.hg.1 |
TC08001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015252.hg.1 |
ENST00000254250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015252.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015252.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015252.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015254.hg.1 |
TC08001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015254.hg.1 |
ENST00000254250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015254.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015254.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015254.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170056.hg.1 |
TC0800010298.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170058.hg.1 |
TC0800010298.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170055.hg.1 |
TC0800010298.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
ENST00000254250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
ENST00000345117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
NM_018105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
NM_199003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170054.hg.1 |
TC0800010298.hg.1 |
Clariom D TC |