Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002028.hg.1 |
ENST00000355819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002028.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002028.hg.1 |
TC12000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002027.hg.1 |
ENST00000355819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002027.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002027.hg.1 |
TC12000156.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147680.hg.1 |
TC1200006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147679.hg.1 |
TC1200006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000355819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000538482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
ENST00000544751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006782.hg.1 |
NM_207345.3 |
Refseq |