Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000498634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000498634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000576562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000576562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
ENST00000576562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000239144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000498634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000498634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000576562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000576562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
ENST00000576562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010996.hg.1 |
NM_024016.3 |
Refseq |