Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002012.hg.1 |
ENST00000313071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002012.hg.1 |
ENST00000382535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002012.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002012.hg.1 |
TC14000184.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006791.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147179.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147179.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147179.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147179.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147179.hg.1 |
TC1400006791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147175.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147175.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147175.hg.1 |
TC1400006791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147178.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147178.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147178.hg.1 |
ENST00000313071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147178.hg.1 |
NM_005249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147178.hg.1 |
TC1400006791.hg.1 |
Clariom D TC |