Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001323668.1 | NP_001310597.1 | 2 | 712 | 114 | |
XM_017023527.1 | XP_016879016.1 | 2 | 885 | 114 | ||
GenBank mRNA | AL832361.1 | - | 2 | 707 | 114 | |
BX647476.1 | - | 2 | 707 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |