Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC007329.2 | - | 3 | 271 | 168 | |
BC068052.1 | - | 3 | 271 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |