Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000472038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000472038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007069.hg.1 |
ENST00000508615.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007069.hg.1 |
ENST00000508615.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000361496.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000367854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000461790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000472038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000472038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000475127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
ENST00000537350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010510.hg.1 |
NM_052862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007069.hg.1 |
ENST00000508615.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007069.hg.1 |
ENST00000508615.2 |
Ensembl |