Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142505.1 | NP_001135977.1 | 3 | 405 | 100 | |
NM_022169.4 | NP_071452.2 | 3 | 629 | 100 | ||
XM_011542952.2 | XP_011541254.1 | 2 | 657 | 100 | ||
XM_017018121.1 | XP_016873610.1 | 3 | 360 | 100 | ||
GenBank mRNA | AJ300465.1 | - | 2 | 292 | 100 | |
AJ308237.1 | - | 2 | 304 | 100 | ||
AJ308239.1 | - | 1 | 63 | 100 | ||
AK289830.1 | - | 3 | 405 | 100 | ||
AK290035.1 | - | 3 | 566 | 100 | ||
BC041091.1 | - | 3 | 629 | 5763981 | 100 | |
DA241043.1 | - | 3 | 405 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |